missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant other TBEV gE Protein

A cDNA sequence encoding the TBEV gE was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP13698-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15944419

  • € 800.00 / 500µg

Estimated Shipment: 30-01-2023
to see stock.
Add to basket

Description

Description

Specifications

Specifications

TBEV gE Protein
500 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Other
Untagged
Protein is >90% pure as determined by SDS- PAGE.
Research Use Only
Recombinant Protein
E. coli
20mM Tris-MES pH 6.5, 8M urea, 200mM NaCl and 0.05% Tween-20.
Videos
SDS
Documents
Certificates
Special Offers

Special Offers