Learn More
enQuireBio™ Recombinant Rat Gremlin 1 / GREM1 Protein
A DNA sequence encoding the Rattus norvegicus (Rat) Gremlin 1 / GREM1, was expressed in the hosts and tags indicated.
€ 245.00 - € 3405.00
Specifications
Accession Number | NP_062155.1 |
---|---|
Gene ID (Entrez) | 50566 |
Name | Gremlin 1 / GREM1 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16090930
|
enQuireBio™
QP6118-EC-100UG |
100 μg |
€ 770.00
100µg |
Estimated Shipment: 01-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16010940
|
enQuireBio™
QP6118-EC-1MG |
1 mg |
€ 3405.00
1mg |
Estimated Shipment: 01-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16000940
|
enQuireBio™
QP6118-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16020940
|
enQuireBio™
QP6118-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16030940
|
enQuireBio™
QP6118-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16040940
|
enQuireBio™
QP6118-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_062155.1 | |
Gremlin 1 / GREM1 Protein | |
Not determined. | |
Full Length | |
Rat | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
50566 | |
Research Use Only | |
Grem1 | |
Recombinant Protein | |
E. coli | |
KKKGSQGAIP PPDKAQHNDS EQTQSPPQPG SRTRGRGQGR GTAMPGEEVL ESSQEALHVT ERKYLKRDWC KTQPLKQTIH EEGCNSRTII NRFCYGQCNS FYIPRHIRKE EGSFQSCSFC KPKKFTTMMV TLNCPELQPP TKKKRVTRVK QCRCISIDLD | |
Greater than 90% as determined by SDS-PAGE. |