missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rhesus Macaque IL 13 Protein
A cDNA sequence encoding the IL 13 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP10691-1mg
Additional Details : Weight : 0.01000kg
Specifications
IL 13 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
SPSPVPRSTA LKELIEELVN ITQNQKAPLC NGSMVWSINL TAGVYCAALE SLINVSGCSA IEKTQRMLNG FCPHKVSAGQ FSSLRVRDTK IEVAQFVKDL LVHLKKLFRE GRFN | |
Greater than 97.0% as determined by SDS-PAGE and HPLC analyses. |
1 mg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Rhesus Macaque | |
Untagged | |
Lyophilized from a 0.2?m filtered concentrated solution in PBS, pH 7.4 containing 5% trehalose. |