missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant S. aureus Serine-aspartate repeat-containing protein 1

A DNA sequence encoding the Staphylococcus aureus (strain Mu50 / ATCC 700699) Serine-aspartate repeat-containing protein E, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7933-ec-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16021691

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

WP_000610237.1
10 μg
Not determined.
Partial (see sequence information for more details).
S. aureus
His
Tris-based buffer, 50% glycerol
S. aureus Serine-aspartate repeat-containing protein 1
Research Use Only
sdrE
Recombinant Protein
E. coli
AENTSTENAK QDDATTSDNK EVVSETENNS TTENDSTNPI KKETNTDSQP EAKEESTTSS TQQQQNNVTA TTETKPQNIE KENVKPSTDK TATEDTSVIL EEKKAPNYTN NDVTTKPSTS EIQTKPTTPQ ESTNIENSQP QPTPSKVDNQ VTDATNPKEP VNVSKEELKN NPEKLKELVR NDNNTDRSTK PVATAPTSVA PKRLNAKMRF AVAQPAAVAS NNVNDLITVT KQTIKVGDGK DNVAAAHDGK DIEYDTEFTI DNKVKKGDTM TINYDKNVIP SDLTDKNDPI DITDPSGEVI AKGTFDKATK QITYTFTDYV DKYEDIKARL TLYSYIDKQA VPNETSLNLT FATAGKETSQ NVSVDYQDPM VHGDSNIQSI FTKLDENKQT IEQQIYVNPL KKTATNTKVD IAGSQVDDYG NIKLGNGSTI IDQNTEIKVY KVNPNQQLPQ SNRIYDFSQY EDVTSQFDNK KSFSNNVATL DFGDINSAYI IKVVSKYTPT SDGELDIAQG TSMRTTDKYG YYNYAGYSNF IVTSNDTGGG DGTVKPEEK
Greater than 90% as determined by SDS-PAGE.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers