missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant S. aureus Serine-aspartate repeat-containing protein 1
A DNA sequence encoding the Staphylococcus aureus (strain Mu50 / ATCC 700699) Serine-aspartate repeat-containing protein E, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7933-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
WP_000610237.1 | |
50 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
S. aureus | |
His | |
Tris-based buffer, 50% glycerol |
S. aureus Serine-aspartate repeat-containing protein 1 | |
Research Use Only | |
sdrE | |
Recombinant Protein | |
E. coli | |
AENTSTENAK QDDATTSDNK EVVSETENNS TTENDSTNPI KKETNTDSQP EAKEESTTSS TQQQQNNVTA TTETKPQNIE KENVKPSTDK TATEDTSVIL EEKKAPNYTN NDVTTKPSTS EIQTKPTTPQ ESTNIENSQP QPTPSKVDNQ VTDATNPKEP VNVSKEELKN NPEKLKELVR NDNNTDRSTK PVATAPTSVA PKRLNAKMRF AVAQPAAVAS NNVNDLITVT KQTIKVGDGK DNVAAAHDGK DIEYDTEFTI DNKVKKGDTM TINYDKNVIP SDLTDKNDPI DITDPSGEVI AKGTFDKATK QITYTFTDYV DKYEDIKARL TLYSYIDKQA VPNETSLNLT FATAGKETSQ NVSVDYQDPM VHGDSNIQSI FTKLDENKQT IEQQIYVNPL KKTATNTKVD IAGSQVDDYG NIKLGNGSTI IDQNTEIKVY KVNPNQQLPQ SNRIYDFSQY EDVTSQFDNK KSFSNNVATL DFGDINSAYI IKVVSKYTPT SDGELDIAQG TSMRTTDKYG YYNYAGYSNF IVTSNDTGGG DGTVKPEEK | |
Greater than 90% as determined by SDS-PAGE. |