missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Toxoplasma Gondii Toxoplasma P32 Protein
A cDNA sequence encoding the Toxoplasma P32 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13788-1mg
Additional Details : Weight : 0.01000kg
Specifications
Toxoplasma Gondii Toxoplasma P32 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
Sterile Filtered solution containing containing PBS and 25mM arginine. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Toxoplasma Gondii | |
His | |
Toxoplasma protein is >95% pure as determined by 10% PAGE (coomassie staining). |