missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Zebrafish d2hgdh, mitochondrial Protein
A DNA sequence encoding the Danio rerio (Zebrafish) (Brachydanio rerio) d2hgdh, mitochondrial, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7418-ec-100ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001074066.1 | |
d2hgdh, mitochondrial Protein | |
Research Use Only | |
d2hgdh | |
Recombinant Protein | |
E. coli | |
ANPANPSAAP PRLPFSRVTQ EDLSFFRALL PGRTITDPDL LKSSNVDWLK TVQGSSDVLL RPKTTEGVSQ ILRYCNERNL AVCPQGGNTG LVGGSVPVFD EIILSTSLMN QVFAFDNISG ILTCQAGCVL ENLSHYLEER DFIMPLDLGA KGSCHIGGNV STNAGGLRLL RYGSLRGTVL GLEVVLADGH VLNCLATLRK DNTGYDLKQL FIGSEGTLGV ITAVSILCPR KPKAVNVAFL GCSSFQQLLE TFQCCRGMLG EILSAFEFLD ASCMNLLEKH LKLTNPITEC PFYIVIETAG SNATHDEEKL HQFLEEVMTS SLVTDGTVAT EATKIKALWS LRERVTEALT HEGYTYKYDI SLPVEKIYDL VQDMRRHLGG MAKNVVGYGH VGDGNLHLNI TSPSKDFDLL AAIEPYVYEW TSQWKGSISA EHGLGLKKRN YIYYSKPSEA VALMGSIKAM LDPKGILNPY KTLPDNIN | |
Greater than 90% as determined by SDS-PAGE. |
565889 | |
100 μg | |
Not determined. | |
Full Length | |
Zebrafish | |
His-SUMO | |
Tris-based buffer, 50% glycerol |