missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Zebrafish d2hgdh, mitochondrial Protein

A DNA sequence encoding the Danio rerio (Zebrafish) (Brachydanio rerio) d2hgdh, mitochondrial, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7418-ec-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16038780

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_001074066.1
d2hgdh, mitochondrial Protein
Research Use Only
d2hgdh
Recombinant Protein
E. coli
ANPANPSAAP PRLPFSRVTQ EDLSFFRALL PGRTITDPDL LKSSNVDWLK TVQGSSDVLL RPKTTEGVSQ ILRYCNERNL AVCPQGGNTG LVGGSVPVFD EIILSTSLMN QVFAFDNISG ILTCQAGCVL ENLSHYLEER DFIMPLDLGA KGSCHIGGNV STNAGGLRLL RYGSLRGTVL GLEVVLADGH VLNCLATLRK DNTGYDLKQL FIGSEGTLGV ITAVSILCPR KPKAVNVAFL GCSSFQQLLE TFQCCRGMLG EILSAFEFLD ASCMNLLEKH LKLTNPITEC PFYIVIETAG SNATHDEEKL HQFLEEVMTS SLVTDGTVAT EATKIKALWS LRERVTEALT HEGYTYKYDI SLPVEKIYDL VQDMRRHLGG MAKNVVGYGH VGDGNLHLNI TSPSKDFDLL AAIEPYVYEW TSQWKGSISA EHGLGLKKRN YIYYSKPSEA VALMGSIKAM LDPKGILNPY KTLPDNIN
Greater than 90% as determined by SDS-PAGE.
565889
10 μg
Not determined.
Full Length
Zebrafish
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers