missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RECQL4 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-95219-0.1ml
This item is not returnable.
View return policy
Description
RECQL4 Polyclonal antibody specifically detects RECQL4 in Human, Mouse samples. It is validated for Western Blot
Specifications
| RECQL4 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| DNA helicase, RecQ-like type 4, DNA helicase, RecQ-like, type 4, EC 3.6.4.12, RecQ protein-like 4RTS, RecQ4, RECQ4ATP-dependent DNA helicase Q4 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 231-330 of human RECQL4 (NP_004251.3). GAGSQGPEASAFQEVSIRVGSPQPSSSGGEKRRWNEEPWESPAQVQQESSQAGPPSEGAGAVAVEEDPPGEPVQAQPPQPCSSPSNPRYHGLSPSSQARA | |
| 0.1 mL | |
| Direct Reversal of DNA Damage | |
| 9401 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction