Learn More
Brady™ Reinforced Polypropylene Escape Route Signs
Brand:  Brady™ 252442
Additional Details : Weight : 0.06900kg
 View more versions of this product
Product Code. 10040854
  • € 13.40 / Each
Estimated Shipment: 23-05-2024
to see stock.
Specifications
Polypropylene
Reinforced Polypropylene
0.369 mm
-40°C to 120°C
Escape Route Sign
1 Ea.
400 mm
200 mm
escape (emergency stairs)
White on Green