Learn More
Brady™ Reinforced Polypropylene Escape Route Signs
Brand: Brady™ 252468
Additional Details : Weight : 0.15100kg
Specifications
Polypropylene | |
Reinforced Polypropylene | |
200 mm | |
200 mm | |
Output | |
White on Green |
1 Ea. | |
First Aid & Emergency exit Sign | |
0.369 mm | |
-40°C to 120°C | |
Escape Route Sign |