missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Replacement Cartridges for PUROKIT Ultrapure Bench Stand Set

Consists of 5 news cartridges and a cleaning kit. Fisherbrand™ Replacement Cartridges for PUROKIT Ultrapure Bench Stand Set does not require any electrical connection.

Brand:  Fisherbrand™

Additional Details : Weight : 0.80000kg

Product Code. 11934199

  • € 364.00 / Each
Estimated Shipment: 21-05-2024
to see stock.
Add to basket

Description

Description

  • Saturation visible by discoloration of R2 columns
  • Change columns easily by unscrewing nuts
  • Shelf life of two years in original packaging at ambient temperature
Specifications

Specifications

R3 Blue cartridge turns to ocher at saturation
30 L/hr.
630 mm
Ultrapure Bench Stand Set PUROKIT
25°C
630 x 170 x 170 mm
Variable as a function of the hardness of the inlet water
Activated Carbon and Ion Exchangers
Water treaatment
2.5 bar
Activated Carbon and Ion Exchangers
60 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers