missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rhodopsin Mouse anti-Amphibian, Avian, Fish, Mammalia, Clone: 4D2, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-59690-100UG
This item is not returnable.
View return policy
Description
Rhodopsin Monoclonal specifically detects Rhodopsin in Amphibian, Avian, Fish, Mammal, Zebrafish samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Frozen.
Specifications
| Rhodopsin | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:1000, ELISA, Immunohistochemistry 1000, Immunocytochemistry/Immunofluorescence, Immunoprecipitation | |
| CSNBAD1, MGC138309, OPN2MGC138311, opsin 2, rod pigment, opsin-2, retinitis pigmentosa 4, autosomal dominant, rhodopsin, RP4 | |
| Mouse | |
| 40 kDa | |
| 100 μg | |
| GPCR, Signal Transduction, Vision | |
| 6010 | |
| Amphibian, Avian, Fish, Mammalia | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
| 4D2 | |
| Unconjugated | |
| PBS pH7.4, 50% glycerol with 0.09% Sodium Azide | |
| RHO | |
| Bovine Rhodopsin | |
| Protein G purified | |
| RUO | |
| Primary | |
| Binds specifically to the N-terminus of Rhodopsin. Does not detect Rhodopsin in invertebrates. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction