missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rhodopsin Mouse anti-Amphibian, Avian, Fish, Mammalia, Clone: 4D2, Novus Biologicals™
Mouse Monoclonal Antibody
€ 383.00 - € 601.00
Specifications
| Antigen | Rhodopsin |
|---|---|
| Clone | 4D2 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:1000, ELISA, Immunohistochemistry 1000, Immunocytochemistry/Immunofluorescence, Immunoprecipitation |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18094506
|
Novus Biologicals
NBP2-59690-25UG |
25 μg |
€ 383.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18058143
|
Novus Biologicals
NBP2-59690-100UG |
100 μg |
€ 601.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Rhodopsin Monoclonal specifically detects Rhodopsin in Amphibian, Avian, Fish, Mammal, Zebrafish samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Frozen.Specifications
| Rhodopsin | |
| 1 mg/mL | |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
| Unconjugated | |
| Mouse | |
| GPCR, Signal Transduction, Vision | |
| PBS pH7.4, 50% glycerol with 0.09% Sodium Azide | |
| 6010 | |
| Bovine Rhodopsin | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| 40 kDa |
| 4D2 | |
| Western Blot 1:1000, ELISA, Immunohistochemistry 1000, Immunocytochemistry/Immunofluorescence, Immunoprecipitation | |
| Monoclonal | |
| Purified | |
| RUO | |
| Amphibian, Avian, Fish, Mammalia | |
| CSNBAD1, MGC138309, OPN2MGC138311, opsin 2, rod pigment, opsin-2, retinitis pigmentosa 4, autosomal dominant, rhodopsin, RP4 | |
| RHO | |
| IgG1 | |
| Protein G purified | |
| Binds specifically to the N-terminus of Rhodopsin. Does not detect Rhodopsin in invertebrates. |
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title