missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ribonuclease A Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-00131-50ul
This item is not returnable.
View return policy
Description
Ribonuclease A Polyclonal antibody specifically detects Ribonuclease A in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| Ribonuclease A | |
| Polyclonal | |
| Immunohistochemistry-Paraffin 1:1000-1:4000 | |
| EC 3.1.27.5, HP-RNase, MGC12408, RIB1, RIB-1, Ribonuclease 1, Ribonuclease A, ribonuclease pancreatic, ribonuclease, RNase A family, 1 (pancreatic), RNase A, RNase UpI-1, RNS1RNase 1 | |
| Produced in rabbits immunized with purified, recombinant Human Ribonuclease A (Accession#: P07998; Met1-Thr156) | |
| 50 μg | |
| DNA replication Transcription Translation and Splicing, Proteases & Other Enzymes | |
| 6035 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction