missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ribonuclease A Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 216.00 - € 326.00
Specifications
| Antigen | Ribonuclease A |
|---|---|
| Dilution | Immunohistochemistry-Paraffin 1:1000-1:4000 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18605486
|
Novus Biologicals
NBP3-00131-100ul |
100 μg |
€ 326.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18637107
|
Novus Biologicals
NBP3-00131-50ul |
50 μg |
€ 216.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Ribonuclease A Polyclonal antibody specifically detects Ribonuclease A in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
| Ribonuclease A | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| DNA replication Transcription Translation and Splicing, Proteases & Other Enzymes | |
| 0.2 um filtered solution in PBS | |
| 6035 | |
| IgG | |
| Antigen and protein A Affinity-purified |
| Immunohistochemistry-Paraffin 1:1000-1:4000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| EC 3.1.27.5, HP-RNase, MGC12408, RIB1, RIB-1, Ribonuclease 1, Ribonuclease A, ribonuclease pancreatic, ribonuclease, RNase A family, 1 (pancreatic), RNase A, RNase UpI-1, RNS1RNase 1 | |
| Produced in rabbits immunized with purified, recombinant Human Ribonuclease A (Accession#: P07998; Met1-Thr156) | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title