missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ribosome maturation protein SBDS Antibody (SR1491), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-22440-50ul
This item is not returnable.
View return policy
Description
Ribosome maturation protein SBDS Monoclonal antibody specifically detects Ribosome maturation protein SBDS in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| Ribosome maturation protein SBDS | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot | |
| SR1491 | |
| Western Blot 1:500-1:2000 | |
| FLJ10917, ribosome maturation protein SBDS, SDSCGI-97, Shwachman-Bodian-Diamond syndrome, Shwachman-Bodian-Diamond syndrome protein, SWDS | |
| A synthesized peptide derived from human Ribosome maturation protein SBDS (Uniprot #: Q9Y3A5) | |
| 50 μg | |
| metabolism | |
| 51119 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction