missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ribosome maturation protein SBDS Antibody (SR1491), Novus Biologicals™
Rabbit Monoclonal Antibody
€ 338.00 - € 449.00
Specifications
| Antigen | Ribosome maturation protein SBDS |
|---|---|
| Clone | SR1491 |
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18683694
|
Novus Biologicals
NBP3-22440-100ul |
100 μg |
€ 449.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18662675
|
Novus Biologicals
NBP3-22440-50ul |
50 μg |
€ 338.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Ribosome maturation protein SBDS Monoclonal antibody specifically detects Ribosome maturation protein SBDS in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| Ribosome maturation protein SBDS | |
| Western Blot 1:500-1:2000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| FLJ10917, ribosome maturation protein SBDS, SDSCGI-97, Shwachman-Bodian-Diamond syndrome, Shwachman-Bodian-Diamond syndrome protein, SWDS | |
| A synthesized peptide derived from human Ribosome maturation protein SBDS (Uniprot #: Q9Y3A5) | |
| Primary | |
| Store at -20° C. Avoid freeze/thaw cycles. |
| SR1491 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| metabolism | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| 51119 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title