missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rictor Antibody (7B3) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 2 publications
Brand: Novus Biologicals NBP1-51645
This item is not returnable.
View return policy
Description
Rictor Monoclonal specifically detects Rictor in Human, Mouse, Primate samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| Rictor | |
| Monoclonal | |
| 1.0 mg/mL | |
| Western Blot 1:500-1:2000, Flow Cytometry 1 ug per million cells, ELISA 1:10000, Immunohistochemistry 1:200-1:1000, Immunocytochemistry/Immunofluorescence 1:200-1:1000, Immunohistochemistry-Paraffin 1:200-1:1000 | |
| AVO3, AVO3 homolog, hAVO3, KIAA1999DKFZp686B11164, mAVO3, MGC39830, PIA, pianissimo, rapamycin-insensitive companion of mTOR, RPTOR independent companion of MTOR, complex 2, TORC2-specific protein AVO3 | |
| Mouse | |
| 192 kDa | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Primate | |
| Precipitation |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 7B3 | |
| Unconjugated | |
| Q6R327 | |
| RICTOR | |
| Purified recombinant fragment of human RICTOR expressed in E. coli. [UniProt# Q6R327] | |
| Ammonium sulfate precipitation | |
| RUO | |
| 253260 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction