missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rictor Antibody (7B3) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 2 publications
€ 231.00 - € 473.00
Specifications
| Antigen | Rictor |
|---|---|
| Clone | 7B3 |
| Concentration | 1.0 mg/mL |
| Dilution | Western Blot 1:500-1:2000, Flow Cytometry 1 ug per million cells, ELISA 1:10000, Immunohistochemistry 1:200-1:1000, Immunocytochemistry/Immunofluorescence 1:200-1:1000, Immunohistochemistry-Paraffin 1:200-1:1000 |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18269264
|
Novus Biologicals
NBP1-51645SS |
0.025 mL |
€ 231.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18069703
|
Novus Biologicals
NBP1-51645 |
0.1 mL |
€ 473.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Rictor Monoclonal specifically detects Rictor in Human, Mouse, Primate samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| Rictor | |
| 1.0 mg/mL | |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Human, Mouse, Primate | |
| AVO3, AVO3 homolog, hAVO3, KIAA1999DKFZp686B11164, mAVO3, MGC39830, PIA, pianissimo, rapamycin-insensitive companion of mTOR, RPTOR independent companion of MTOR, complex 2, TORC2-specific protein AVO3 | |
| RICTOR | |
| IgG1 | |
| Ammonium sulfate precipitation | |
| 192 kDa |
| 7B3 | |
| Western Blot 1:500-1:2000, Flow Cytometry 1 ug per million cells, ELISA 1:10000, Immunohistochemistry 1:200-1:1000, Immunocytochemistry/Immunofluorescence 1:200-1:1000, Immunohistochemistry-Paraffin 1:200-1:1000 | |
| Monoclonal | |
| Precipitation | |
| RUO | |
| Q6R327 | |
| 253260 | |
| Purified recombinant fragment of human RICTOR expressed in E. coli. [UniProt# Q6R327] | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title