missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RIPK3/RIP3, p Ser232 Antibody (SR1864), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-22611-100ul
This item is not returnable.
View return policy
Description
RIPK3/RIP3 Monoclonal antibody specifically detects RIPK3/RIP3 in Mouse samples. It is validated for Western Blot
Specifications
| RIPK3/RIP3 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
| Western Blot | |
| SR1864 | |
| Western Blot 1:500-1:2000 | |
| EC 2.7.11.1, Receptor Interacting Serine/Threonine Kinase 3, Receptor-interacting protein 3, receptor-interacting serine/threonine-protein kinase 3, receptor-interacting serine-threonine kinase 3, RIP3, RIP-3, RIP-like protein kinase 3 | |
| A synthesized peptide derived from human phospho-RIPK3/RIP3 [Ser232] (Uniprot #: Q9Y572, Q9QZL0) | |
| 100 μg | |
| Cancer, Cell Biology, Immunology, Necroptosis, Protein Kinase, Signal Transduction | |
| 11035 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction