missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RIPK3/RIP3, p Ser232 Antibody (SR1864), Novus Biologicals™
Rabbit Monoclonal Antibody
€ 349.00 - € 472.00
Specifications
| Antigen | RIPK3/RIP3 |
|---|---|
| Clone | SR1864 |
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18615274
|
Novus Biologicals
NBP3-22611-100ul |
100 μg |
€ 472.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18636454
|
Novus Biologicals
NBP3-22611-50ul |
50 μg |
€ 349.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
RIPK3/RIP3 Monoclonal antibody specifically detects RIPK3/RIP3 in Mouse samples. It is validated for Western BlotSpecifications
| RIPK3/RIP3 | |
| Western Blot 1:500-1:2000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Mouse | |
| EC 2.7.11.1, Receptor Interacting Serine/Threonine Kinase 3, Receptor-interacting protein 3, receptor-interacting serine/threonine-protein kinase 3, receptor-interacting serine-threonine kinase 3, RIP3, RIP-3, RIP-like protein kinase 3 | |
| A synthesized peptide derived from human phospho-RIPK3/RIP3 [Ser232] (Uniprot #: Q9Y572, Q9QZL0) | |
| Primary | |
| Store at -20° C. Avoid freeze/thaw cycles. |
| SR1864 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cancer, Cell Biology, Immunology, Necroptosis, Protein Kinase, Signal Transduction | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| 11035 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title