missing translation for 'onlineSavingsMsg'
Learn More

RIPK3/RIP3, p Ser232 Antibody (SR1864), Novus Biologicals™

Rabbit Monoclonal Antibody

Brand:  Novus Biologicals NBP3-22611-50ul

 View more versions of this product

Product Code. 18636454

  • € 349.00 / 50µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

RIPK3/RIP3 Monoclonal antibody specifically detects RIPK3/RIP3 in Mouse samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Specifications

Specifications

RIPK3/RIP3
Monoclonal
Unconjugated
PBS, pH 7.4, 150mM NaCl, 50% glycerol.
Rabbit
Affinity purified
RUO
Primary
Mouse
Purified
Western Blot
SR1864
Western Blot 1:500-1:2000
EC 2.7.11.1, Receptor Interacting Serine/Threonine Kinase 3, Receptor-interacting protein 3, receptor-interacting serine/threonine-protein kinase 3, receptor-interacting serine-threonine kinase 3, RIP3, RIP-3, RIP-like protein kinase 3
A synthesized peptide derived from human phospho-RIPK3/RIP3 [Ser232] (Uniprot #: Q9Y572, Q9QZL0)
50 μg
Cancer, Cell Biology, Immunology, Necroptosis, Protein Kinase, Signal Transduction
11035
Store at -20° C. Avoid freeze/thaw cycles.
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.