Learn More
RIT2 Monoclonal Antibody (OTI4B5), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA501704
This item is not returnable.
View return policy
Description
RIT2 belongs to the RAS (HRAS) superfamily of small GTPases.Specifications
| RIT2 | |
| Monoclonal | |
| 0.84 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q99578 | |
| RIT2 | |
| Full length human recombit protein of human RIT2 produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot | |
| OTI4B5 | |
| Unconjugated | |
| RIT2 | |
| RIBA, RIN, ROC2 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 6014 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |