missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RMND5A Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94019-0.02ml
This item is not returnable.
View return policy
Description
RMND5A Polyclonal antibody specifically detects RMND5A in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| RMND5A | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| C-terminal to LisH motif, 44 kDa, CTLH, FLJ12753, FLJ13910, FLJ21795, MGC78451, p44CTLH, protein RMD5 homolog A, required for meiotic nuclear division 5 homolog A (S. cerevisiae), RMD5 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 200-270 of human RMND5A (NP_073617.1). ISLLMGGTTNQREALQYAKNFQPFALNHQKDIQVLMGSLVYLRQGIENSPYVHLLDANQWADICDIFTRDA | |
| 0.02 mL | |
| Stem Cell Markers | |
| 64795 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction