missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RNASE3 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93655-0.02ml
This item is not returnable.
View return policy
Description
RNASE3 Polyclonal antibody specifically detects RNASE3 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| RNASE3 | |
| Polyclonal | |
| Immunohistochemistry 1:50-1:100, Immunohistochemistry-Paraffin | |
| eosinophil cationic protein, ECP, ribonuclease, RNase A family, 3, RNS3 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 28-160 of human RNASE3 (NP_002926.2). RPPQFTRAQWFAIQHISLNPPRCTIAMRAINNYRWRCKNQNTFLRTTFANVVNVCGNQSIRCPHNRTLNNCHRSRFRVPLLHCDLINPGAQNISNCTYADRPGRRFYVVACDNRDPRDSPRYPVVPVHLDTTI | |
| 0.02 mL | |
| Proteases & Other Enzymes | |
| 6037 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction