missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RNASE3 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 190.00 - € 470.00
Specifications
| Antigen | RNASE3 |
|---|---|
| Dilution | Immunohistochemistry 1:50-1:100, Immunohistochemistry-Paraffin |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18630681
|
Novus Biologicals
NBP2-93655-0.02ml |
0.02 mL |
€ 190.00
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18620781
|
Novus Biologicals
NBP2-93655-0.1ml |
0.1 mL |
€ 470.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
RNASE3 Polyclonal antibody specifically detects RNASE3 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| RNASE3 | |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Proteases & Other Enzymes | |
| PBS (pH 7.3), 50% glycerol | |
| 6037 | |
| IgG | |
| Affinity purified |
| Immunohistochemistry 1:50-1:100, Immunohistochemistry-Paraffin | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| eosinophil cationic protein, ECP, ribonuclease, RNase A family, 3, RNS3 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 28-160 of human RNASE3 (NP_002926.2). RPPQFTRAQWFAIQHISLNPPRCTIAMRAINNYRWRCKNQNTFLRTTFANVVNVCGNQSIRCPHNRTLNNCHRSRFRVPLLHCDLINPGAQNISNCTYADRPGRRFYVVACDNRDPRDSPRYPVVPVHLDTTI | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title