missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RNF150 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94857-0.02ml
This item is not returnable.
View return policy
Description
RNF150 Polyclonal antibody specifically detects RNF150 in Mouse, Rat samples. It is validated for Western Blot
Specifications
| RNF150 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| KIAA1214, MGC125502, ring finger protein 150 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 92-211 of human RNF150 (NP_065775.1). MASSAHDRLACDPNTKFAAPTRGKNWIALIPKGNCTYRDKIRNAFLQNASAVVIFNVGSNTNETITMPHAGVEDIVAIMIPEPKGKEIVSLLERNITVTMYITIGTRNLQKYVSRTSVVF | |
| 0.02 mL | |
| Zinc Finger | |
| 57484 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction