missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Ro 3306

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
Ro 3306 is an ATP-competitive cyclin-dependent kinase 1 (cdk1) inhibitor (Ki values range from 35 to 240 nM depending on cdk1 binding partner). Also reported to inhibit other cdks (Ki values are 0.89-1.32, 0.03 and >2 μM for cdk2, cdk3 and cdk4, respectively). Induces G2/M phase cell cycle arrest and apoptosis. Downregulates the expression of antiapoptotic proteins such as Bcl-2 and survivin and enhances downstream p53 signaling in acute myeloid leukemia (AML). RO 3306 also improves homology-directed repair (HDR) -mediated gene editing in hematopoietic stem and progenitor cells.
Specifications
Tekniske data
| Quantity | 50 mg |
| Formula Weight | 351.45 |
| Purity | >98% |
| Molecular Weight (g/mol) | 351.442 |
| Chemical Name or Material | Ro 3306 |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion