missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rocuronium Bromide, MedChemExpress
Description
Rocuronium Bromide (ORG 9426 Bromide) is an aminosteroid non-depolarizing neuromuscular blocker or muscle relaxant used in modern anaesthesia, to facilitate endotracheal intubation and to provide skeletal musclerelaxation during surgery or mechanical ventilation.
Specifications
Specifications
| Chemical Name or Material | Rocuronium Bromide |
| CAS | 119302-91-9 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C32H53BrN2O4 |
| Quantity | 10 mg |
| Synonym | ORG 9426 Bromide |
| Solubility Information | DMSO : 100 mg/mL (164.02 mM; Need ultrasonic) ∣H2O : 100 mg/mL (164.02 mM; Need ultrasonic) |
| SMILES | C[C@@]12[C@](C[C@H]([N+]3(CC=C)CCCC3)[C@@H]2OC(C)=O)([H])[C@]([C@]4([H])CC1)([H])CC[C@@]([C@]4(C)C5)([H])C[C@H](O)[C@H]5N6CCOCC6.[Br-] |
| Molecular Weight (g/mol) | 609.68 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?