missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ROR2 Antibody (SR1037), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-22166-50ul
This item is not returnable.
View return policy
Description
ROR2 Monoclonal antibody specifically detects ROR2 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunofluorescence
Specifications
| ROR2 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry, Immunofluorescence | |
| SR1037 | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:50, Immunocytochemistry/Immunofluorescence 1:50-1:200 | |
| BDB, BDB1, EC 2.7.10.1, MGC163394, neurotrophic tyrosine kinase receptor-related 2, Neurotrophic tyrosine kinase, receptor-related 2, NTRKR2tyrosine-protein kinase transmembrane receptor ROR2, receptor tyrosine kinase-like orphan receptor 2 | |
| A synthesized peptide derived from human ROR2 (Uniprot #: Q01974) | |
| 50 μg | |
| Protein Kinase, Wnt Signaling Pathway | |
| 4920 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction