missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ROR2 Antibody (SR1037), Novus Biologicals™
Rabbit Monoclonal Antibody
€ 338.00 - € 449.00
Specifications
| Antigen | ROR2 |
|---|---|
| Clone | SR1037 |
| Dilution | Western Blot 1:500-1:2000, Flow Cytometry 1:50, Immunocytochemistry/Immunofluorescence 1:50-1:200 |
| Applications | Western Blot, Flow Cytometry, Immunofluorescence |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18672835
|
Novus Biologicals
NBP3-22166-100ul |
100 μg |
€ 449.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18692664
|
Novus Biologicals
NBP3-22166-50ul |
50 μg |
€ 338.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ROR2 Monoclonal antibody specifically detects ROR2 in Human samples. It is validated for Western Blot, Flow Cytometry, ImmunofluorescenceSpecifications
| ROR2 | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:50, Immunocytochemistry/Immunofluorescence 1:50-1:200 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| BDB, BDB1, EC 2.7.10.1, MGC163394, neurotrophic tyrosine kinase receptor-related 2, Neurotrophic tyrosine kinase, receptor-related 2, NTRKR2tyrosine-protein kinase transmembrane receptor ROR2, receptor tyrosine kinase-like orphan receptor 2 | |
| A synthesized peptide derived from human ROR2 (Uniprot #: Q01974) | |
| Primary | |
| Store at -20° C. Avoid freeze/thaw cycles. |
| SR1037 | |
| Western Blot, Flow Cytometry, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Protein Kinase, Wnt Signaling Pathway | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| 4920 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title