missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RORB Monoclonal Antibody (OTI3D7), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA806860S
This item is not returnable.
View return policy
Description
RORB Monoclonal antibody specifically detects RORB in Human samples. It is validated for Western BlotSpecifications
| RORB | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q92753 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 6096 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG3 |
| Western Blot | |
| OTI3D7 | |
| Unconjugated | |
| RORB | |
| bA133M9.1; Nr1f2; Nuclear receptor ROR-beta; nuclear receptor RZR-beta; Nuclear receptor subfamily 1 group F member 2; RAR related orphan receptor B; RAR-related orphan receptor B; RAR-related orphan receptor beta; retinoic acid-binding receptor beta; retinoid-related orphan receptor b; retinoid-related orphan receptor beta; retinoid-related orphan receptor-beta; ror beta; Rorb; Rorbeta; ROR-BETA; rzrb; RZR-beta; sb:eu943; transcription factor; Unknown (protein for IMAGE:7173671) | |
| Human recombinant protein fragment corresponding to amino acids 1-260 of human RORB produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction