missing translation for 'onlineSavingsMsg'
Learn More

RORB Monoclonal Antibody (OTI3D7), TrueMAB™, OriGene

Mouse Monoclonal Antibody

POA

Specifications

Antigen RORB
Clone OTI3D7
Applications Western Blot
Classification Monoclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
17567143
Encompass_Preferred
View Documents
Origene Technologies
TA806860S
30 μL
POA
30µL
View alternative products
Save Up to 
»
N/A
Please to purchase this item. Need a web account? Register with us today!
This item requires a price update prior to being ordered.

Request a Price for this item here.

17547143
Encompass_Preferred
View Documents
Origene Technologies
TA806860
100 μL
POA
100µL
View alternative products
Save Up to 
»
N/A
Please to purchase this item. Need a web account? Register with us today!
This item requires a price update prior to being ordered.

Request a Price for this item here.

Description

Description

RORB Monoclonal antibody specifically detects RORB in Human samples. It is validated for Western Blot
Specifications

Specifications

RORB
Western Blot
Unconjugated
Mouse
Human
6096
Primary
RORB
OTI3D7
Monoclonal
Liquid
RUO
Q92753
IgG3
-20° C, Avoid Freeze/Thaw Cycles
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.