Learn More
RORB Monoclonal Antibody (OTI3D7), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA806860
Encompass_Preferred
 View more versions of this product
Product Code. 17547143
  • POA / 100µL
This item requires a price update prior to being ordered.
Request a Price for this item here.
View alternative products »
Description
Description
Retinoid-related orphan receptor beta (ROR beta), a NR1 thyroid hormone receptor, affects the development and functioning of the central nervous system, including sensory input integration and circadian timing. Disruption of ROR beta in mice results in juvenile ataxia, retinal degeneration, circadian activity abnormalities, and delayed onset of male fertility. ROR beta binds to the monomeric response elements containing the sequence AnnTAGGTCA as a monomer. An alternatively spliced isoform, ROR beta2, has been isolated from rats, and the expression of ROR beta2 is confined to the pineal gland and retina. ROR beta2 shares common DNA- and putative ligand-binding domains with wild-type ROR beta but is characterized by a different amino-terminal domain. ROR beta expression has been documented in various regions of the mouse brain.
Specifications
Specifications
RORB
Monoclonal
Unconjugated
RORB
NR1F2, ROR-BETA, RZR-BETA, RZRB, bA133M9.1
Mouse
Affinity Chromatography
RUO
6096
-20° C, Avoid Freeze/Thaw Cycles
Liquid
Western Blot
OTI3D7
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q92753
RORB
Human recombit protein fragment corresponding to amino acids 1-260 of human RORB produced in E.coli.
100 μL
Primary
Human
Antibody
IgG3