missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ROS Antibody (7-2B) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50182
This item is not returnable.
View return policy
Description
ROS Monoclonal antibody specifically detects ROS in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Functional Assay
Specifications
| ROS | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:100 - 1:2000, Immunocytochemistry/ Immunofluorescence 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500, Functional | |
| proto-oncogene tyrosine-protein kinase ROS, c-ros oncogene 1, receptor tyrosine kinase, c-ros-1, MCF3, ROS | |
| Extracellular portion of ROS amino acid 1-285 fused to Fc, transiently expressed in 293 cells and purified using PtnA column chromatography. | |
| 0.1 mL | |
| Protein Kinase | |
| 6098 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
| Western Blot, Immunofluorescence, Immunoprecipitation, Functional Assay | |
| 7-2B | |
| Unconjugated | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction