missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ROS Antibody (7-2B) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
€ 190.00 - € 351.00
Specifications
| Antigen | ROS |
|---|---|
| Clone | 7-2B |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:100 - 1:2000, Immunocytochemistry/ Immunofluorescence 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500, Functional |
| Applications | Western Blot, Immunofluorescence, Immunoprecipitation, Functional Assay |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18655728
|
Novus Biologicals
NBP2-50182-0.025ml |
0.025 mL |
€ 190.00
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18672278
|
Novus Biologicals
NBP2-50182 |
0.1 mL |
€ 351.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ROS Monoclonal antibody specifically detects ROS in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Functional AssaySpecifications
| ROS | |
| 1 mg/mL | |
| Western Blot, Immunofluorescence, Immunoprecipitation, Functional Assay | |
| Unconjugated | |
| Mouse | |
| Protein Kinase | |
| PBS | |
| 6098 | |
| IgG1 | |
| Protein A purified |
| 7-2B | |
| Western Blot 1:100 - 1:2000, Immunocytochemistry/ Immunofluorescence 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500, Functional | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| proto-oncogene tyrosine-protein kinase ROS, c-ros oncogene 1, receptor tyrosine kinase, c-ros-1, MCF3, ROS | |
| Extracellular portion of ROS amino acid 1-285 fused to Fc, transiently expressed in 293 cells and purified using PtnA column chromatography. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title