missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RPA2, p Thr21 Antibody (3B2), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-26396-50ul
This item is not returnable.
View return policy
Description
RPA2 Monoclonal antibody specifically detects RPA2 in Human samples. It is validated for ELISA, Immunocytochemistry
Specifications
| RPA2 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA, Immunocytochemistry | |
| 3B2 | |
| ELISA, Immunocytochemistry/Immunofluorescence 1:20-1:200 | |
| REPA2, Replication factor A protein 2, replication protein A 32 kDa subunit, Replication protein A 34 kDa subunit, replication protein A2 (32kD), replication protein A2, 32kDa, RF-A protein 2, RP-A p32, RP-A p34, RPA32, RPA34 | |
| A synthesized peptide derived from Human RPA2 [p Thr21] [UniProt P15927] | |
| 50 μL | |
| Cancer, Cell Cycle and Replication, Checkpoint signaling, Core ESC Like Genes, DNA Double Strand Break Repair, DNA Repair, Nucleotide Excision Repair, Phospho Specific, Stem Cell Markers | |
| 6118 | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction