missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RPA2, p Thr21 Antibody (3B2), Novus Biologicals™
Rabbit Monoclonal Antibody
€ 343.00 - € 533.00
Specifications
| Antigen | RPA2 |
|---|---|
| Clone | 3B2 |
| Dilution | ELISA, Immunocytochemistry/Immunofluorescence 1:20-1:200 |
| Applications | ELISA, Immunocytochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30025318
|
Novus Biologicals
NBP3-26396-50ul |
50 μL |
€ 343.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30024794
|
Novus Biologicals
NBP3-26396-100ul |
100 μL |
€ 533.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
RPA2 Monoclonal antibody specifically detects RPA2 in Human samples. It is validated for ELISA, ImmunocytochemistrySpecifications
| RPA2 | |
| ELISA, Immunocytochemistry/Immunofluorescence 1:20-1:200 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| REPA2, Replication factor A protein 2, replication protein A 32 kDa subunit, Replication protein A 34 kDa subunit, replication protein A2 (32kD), replication protein A2, 32kDa, RF-A protein 2, RP-A p32, RP-A p34, RPA32, RPA34 | |
| A synthesized peptide derived from Human RPA2 [p Thr21] [UniProt P15927] | |
| Primary | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. |
| 3B2 | |
| ELISA, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Cancer, Cell Cycle and Replication, Checkpoint signaling, Core ESC Like Genes, DNA Double Strand Break Repair, DNA Repair, Nucleotide Excision Repair, Phospho Specific, Stem Cell Markers | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| 6118 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title