missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RPL22 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93082-0.02ml
This item is not returnable.
View return policy
Description
RPL22 Polyclonal antibody specifically detects RPL22 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Gene Knock-Out
Specifications
| RPL22 | |
| Polyclonal | |
| Western Blot 1:1000 - 1:5000, Immunohistochemistry 1:50 - 1:100, Immunohistochemistry-Paraffin, Knockout Validated | |
| EAPHBP15, EBER-associated protein, Epstein-Barr virus small RNA-associated protein, Epstein-Barr-encoded RNA-associated protein, HBP15/L22, heparin-binding protein 15, Heparin-binding protein HBp15, ribosomal protein L22,60S ribosomal protein L22 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-122 of human RPL22 (NP_000974.1). MAPVKKLVVKGGKKKKQVLKFTLDCTHPVEDGIMDAANFEQFLQERIKVNGKAGNLGGGVVTIERSKSKITVTSEVPFSKRYLKYLTKKYLKKNNLRDWLRVVANSKESYELRYFQINQDEE | |
| 0.02 mL | |
| Core ESC Like Genes, Stem Cell Markers | |
| 6146 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Gene Knock-Out | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction