missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RPL22 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 209.00 - € 471.00
Specifications
| Antigen | RPL22 |
|---|---|
| Dilution | Western Blot 1:1000 - 1:5000, Immunohistochemistry 1:50 - 1:100, Immunohistochemistry-Paraffin, Knockout Validated |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Gene Knock-Out |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18625910
|
Novus Biologicals
NBP2-93082-0.02ml |
0.02 mL |
€ 209.00
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18634651
|
Novus Biologicals
NBP2-93082-0.1ml |
0.1 mL |
€ 471.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
RPL22 Polyclonal antibody specifically detects RPL22 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Gene Knock-OutSpecifications
| RPL22 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Gene Knock-Out | |
| Unconjugated | |
| Rabbit | |
| Core ESC Like Genes, Stem Cell Markers | |
| PBS (pH 7.3), 50% glycerol | |
| 6146 | |
| IgG | |
| Affinity purified |
| Western Blot 1:1000 - 1:5000, Immunohistochemistry 1:50 - 1:100, Immunohistochemistry-Paraffin, Knockout Validated | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| EAPHBP15, EBER-associated protein, Epstein-Barr virus small RNA-associated protein, Epstein-Barr-encoded RNA-associated protein, HBP15/L22, heparin-binding protein 15, Heparin-binding protein HBp15, ribosomal protein L22,60S ribosomal protein L22 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-122 of human RPL22 (NP_000974.1). MAPVKKLVVKGGKKKKQVLKFTLDCTHPVEDGIMDAANFEQFLQERIKVNGKAGNLGGGVVTIERSKSKITVTSEVPFSKRYLKYLTKKYLKKNNLRDWLRVVANSKESYELRYFQINQDEE | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title