missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RRAGD Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93833-0.02ml
This item is not returnable.
View return policy
Description
RRAGD Polyclonal antibody specifically detects RRAGD in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| RRAGD | |
| Polyclonal | |
| Western Blot 1:1000-1:4000, Immunohistochemistry 1:50-1:200, Immunohistochemistry-Paraffin | |
| bA11D8.2.1, DKFZP761H171, FLJ44176, Rag D, Rag D protein, RAGD, Ras-related GTP binding D, ras-related GTP-binding protein D | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 181-400 of human RRAGD (NP_067067.1). VDGLSDDHKIETQRDIHQRANDDLADAGLEKIHLSFYLTSIYDHSIFEAFSKVVQKLIPQLPTLENLLNIFISNSGIEKAFLFDVVSKIYIATDSTPVDMQTYELCCDMIDVVIDISCIYGLKEDGAGTPYDKESTAIIKLNNTTVLYLKEVTKFLALVCFVREESFERKGLIDYNFHCFRKAIHEVFEVRMKVVKSRKVQNRLQKKKRATPNGTPRVLL | |
| 0.02 mL | |
| Cancer, mTOR Pathway | |
| 58528 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction