missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RRAGD Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 214.00 - € 493.00
Specifications
| Antigen | RRAGD |
|---|---|
| Dilution | Western Blot 1:1000-1:4000, Immunohistochemistry 1:50-1:200, Immunohistochemistry-Paraffin |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18612711
|
Novus Biologicals
NBP2-93833-0.02ml |
0.02 mL |
€ 214.00
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18676540
|
Novus Biologicals
NBP2-93833-0.1ml |
0.1 mL |
€ 493.00
0.01mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
RRAGD Polyclonal antibody specifically detects RRAGD in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| RRAGD | |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Cancer, mTOR Pathway | |
| PBS (pH 7.3), 50% glycerol | |
| 58528 | |
| IgG | |
| Affinity purified |
| Western Blot 1:1000-1:4000, Immunohistochemistry 1:50-1:200, Immunohistochemistry-Paraffin | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| bA11D8.2.1, DKFZP761H171, FLJ44176, Rag D, Rag D protein, RAGD, Ras-related GTP binding D, ras-related GTP-binding protein D | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 181-400 of human RRAGD (NP_067067.1). VDGLSDDHKIETQRDIHQRANDDLADAGLEKIHLSFYLTSIYDHSIFEAFSKVVQKLIPQLPTLENLLNIFISNSGIEKAFLFDVVSKIYIATDSTPVDMQTYELCCDMIDVVIDISCIYGLKEDGAGTPYDKESTAIIKLNNTTVLYLKEVTKFLALVCFVREESFERKGLIDYNFHCFRKAIHEVFEVRMKVVKSRKVQNRLQKKKRATPNGTPRVLL | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title