missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RT1.B Monoclonal Antibody (OX-6)
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA517427
This item is not returnable.
View return policy
Description
RT1.B Monoclonal Antibody for Western Blot, IHC (F), IHC (P), Flow
gene complex that may play a role in antigen presentation.
Specifications
| RT1.B | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 0.02% sodium azide | |
| 0 | |
| RT1-B | |
| Rat thymocyte membrane glycoprotein. | |
| 500 μg | |
| Primary | |
| Mouse, Rat | |
| Antibody | |
| IgG1 |
| Flow Cytometry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot | |
| OX-6 | |
| Unconjugated | |
| RT1-B | |
| rat class I B; RT1 class II, locus B; RT1-B | |
| Mouse | |
| Protein G | |
| RUO | |
| 24738 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction