missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RT1.B Monoclonal Antibody (OX-6)

Mouse Monoclonal Antibody
€ 367.00 - € 582.00
Specifications
| Antigen | RT1.B |
|---|---|
| Clone | OX-6 |
| Concentration | 1 mg/mL |
| Content And Storage | Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
| Applications | Flow Cytometry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11544403
![]() |
Invitrogen™
MA170008 |
250 μg |
€ 367.00
250µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15358330
|
Invitrogen™
MA517427 |
500 μg |
€ 582.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
RT1.B Monoclonal Antibody for Western Blot, IHC (F), IHC (P), Flow
gene complex that may play a role in antigen presentation.Specifications
| RT1.B | |
| 1 mg/mL | |
| Flow Cytometry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Mouse | |
| RUO | |
| PBS with 0.02% sodium azide | |
| 0 | |
| 24738 | |
| Rat thymocyte membrane glycoprotein. | |
| Antibody |
| OX-6 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Monoclonal | |
| Liquid | |
| IgG1 | |
| Mouse, Rat | |
| RT1-B | |
| rat class I B; RT1 class II, locus B; RT1-B | |
| RT1-B | |
| Primary | |
| Protein G |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title