missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RUSC1 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93950-0.02ml
This item is not returnable.
View return policy
Description
RUSC1 Polyclonal antibody specifically detects RUSC1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence
Specifications
| RUSC1 | |
| Polyclonal | |
| Western Blot 1:200 - 1:2000, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence 1:50 - 1:200 | |
| DKFZp761A1822, Nesca, NESCARUN and SH3 domain-containing protein 1, New molecule containing SH3 at the carboxy-terminus, RUN and SH3 domain containing 1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-320 of human RUSC1 (NP_055143.2). MAEAQSGTGQLQEQKKGLLIAVSVSVDKIISHFGAARNLVQKAQLGDSRLSPDVGHLVLTTLCPALHALVADGLKPFRKDLITGQRRSSPWSVVEASVKPGSSTRSLGTLYSQVSRLAPLSSSRSRFHAFILGLLNTKQLELWFSSLQEDAGLLSLLYLPTGFFSLARGGCPSLSTELLLLLQPLSVLTFHLDLLFEHHHHLPLGPPQAPAPPGPPPALQQTMQAMLHFGGRLAQSLRGTSKEAASDPSDSPNLPTPGSWWEQLTQASRVYASGGTEGFPLSRWAPGRHGTAAEEGAQERPLPTDEMAPGRGLWLGRLFG | |
| 0.02 mL | |
| Neuroscience, Signal Transduction | |
| 23623 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction