missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RuvA Antibody (RuvA 12C6) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50278
This item is not returnable.
View return policy
Description
RuvA Monoclonal antibody specifically detects RuvA in E. coli samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Radioimmune Assays
Specifications
| RuvA | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500, Radioimmunoassay 1:100 - 1:2000 | |
| EC 3.1.22.4, Holliday junction ATP dependent DNA helicase ruvA, Ruv A | |
| Ruv A protein from E. Coli | |
| 0.1 mL | |
| Epigenetics | |
| E. coli | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Radioimmune Assays (RIA) | |
| RuvA 12C6 | |
| Unconjugated | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction