missing translation for 'onlineSavingsMsg'
Learn More

RuvA Antibody (RuvA 12C6) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody

€ 190.00 - € 351.00

Specifications

Antigen RuvA
Clone RuvA 12C6
Concentration 1 mg/mL
Dilution Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500, Radioimmunoassay 1:100 - 1:2000
Applications Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Radioimmune Assays (RIA)
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18677247
View Documents
Novus Biologicals
NBP2-50278-0.025ml
0.025 mL
€ 190.00
0.03mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18644497
View Documents
Novus Biologicals
NBP2-50278
0.1 mL
€ 351.00
0.10mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

RuvA Monoclonal antibody specifically detects RuvA in E. coli samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Radioimmune Assays
Specifications

Specifications

RuvA
1 mg/mL
Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Radioimmune Assays (RIA)
Unconjugated
Mouse
Epigenetics
PBS
Ruv A protein from E. Coli
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
RuvA 12C6
Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500, Radioimmunoassay 1:100 - 1:2000
Monoclonal
Purified
RUO
E. coli
EC 3.1.22.4, Holliday junction ATP dependent DNA helicase ruvA, Ruv A
IgG1
Protein A purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.