Learn More
Novus Biologicals™ Ryk Antibody (F35 P7 D7 F5), Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals™ NBP3-30896-200ug
 View more versions of this product
Product Code. 30066879
  • € 508.00 / 200µg
Description
Ryk Monoclonal antibody specifically detects Ryk in Human,Mouse,Rat samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Ryk
Monoclonal
Unconjugated
Phosphate buffered saline
Mouse
Protein A or G purified
RUO
Primary
Human, Mouse, Rat
Purified
Western Blot
F35 P7 D7 F5
Western Blot : 1-5 μg/mL
D3S3195, EC 2.7.10, EC 2.7.10.1, hydroxyaryl-protein kinase, JTK5, JTK5A, JTK5A protein tyrosine kinase, RYK receptor-like tyrosine kinase, RYK1, tyrosine-protein kinase RYK
Hybridoma produced by the fusion of splenocytes from BALB/c mice immunized with a synthetic peptide derived from the human Ryk protein and mouse myeloma Ag8563 cells. Sequence common in human, mouse and rat.
200 μg
Protein Kinase
6259
Store at -20°C. Avoid freeze/thaw cycles.