missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-1,2-Propanediol, 97%

Product Code. p-7026005
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11490397

Brand: Thermo Scientific Alfa Aesar B21773.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 4254-15-3 | C3H8O2 | 76.10 g/mol

(S)-(+)-1,2-Propanediol acts as an organic solvent and diluent used in pharmaceutical preparations. It is also used as a chiral synthetic intermediate and substrate for enzyme studies.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(S)-(+)-1,2-Propanediol acts as an organic solvent and diluent used in pharmaceutical preparations. It is also used as a chiral synthetic intermediate and substrate for enzyme studies.

Solubility
Miscible with water.

Notes
Hygroscopic. Incompatible with strong oxidizing agents, acid chlorides, acid anhydrides, oxidizing agents,chloroformates and reducing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4254-15-3
Molecular Formula C3H8O2
Molecular Weight (g/mol) 76.10
MDL Number MFCD00004539
InChI Key DNIAPMSPPWPWGF-UHFFFAOYNA-N
Synonym s-+-1,2-propanediol, s-propane-1,2-diol, s-1,2-propanediol, 2s-propane-1,2-diol, s-propylene glycol, s-+-propylene glycol, s-+-1,2-dihydroxypropane, propylene glycol #, pgo
PubChem CID 439846
ChEBI CHEBI:29002
SMILES CC(O)CO

Specifications

Density 1.04
Boiling Point 186°C to 188°C
Flash Point 107°C (224°F)
Refractive Index 1.432
Quantity 1 g
Beilstein 1718871
Sensitivity Hygroscopic
Merck Index 14,7855
Solubility Information Miscible with water.
Optical Rotation +16° (Neat)
IUPAC Name (2S)-propane-1,2-diol
Formula Weight 76.1
Percent Purity 97%
Chemical Name or Material (S)-(+)-1,2-Propanediol
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.